Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kgb-1
Entrez Gene ID
176866
Gene Name
GLH-binding kinase 1
Gene Aliases
CELE_T07A9.3
Chromosome Location
Chr.IV: 405622 - 410035 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067521.7 NP_499922.1 8 - 9 1014 153

View Full Product Details