Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glb-2
Entrez Gene ID
182330
Gene Name
GLoBin related
Gene Aliases
CELE_C06E4.7
Chromosome Location
Chr.IV: 7262032 - 7263237 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068756.3 NP_501157.2 2 - 3 201 73

View Full Product Details