Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rod-1
Entrez Gene ID
177520
Gene Name
Kinetochore-associated protein rod-1
Gene Aliases
CELE_F55G1.4
Chromosome Location
Chr.IV: 7478171 - 7485527 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068799.7 NP_501200.1 13 - 14 4651 133

View Full Product Details