Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
itr-1
Entrez Gene ID
177546
Gene Name
protein-coding
Gene Aliases
CELE_F33D4.2
Chromosome Location
Chr.IV: 7675174 - 7696930 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027999.4 NP_001023170.1 13 - 14 2251 63
NM_001028000.5 NP_001023171.1 11 - 12 2083 63
NM_001028001.5 NP_001023172.1 13 - 14 2208 63
NM_001028002.4 NP_001023173.1 12 - 13 2179 63
NM_001268376.4 NP_001255305.1 12 - 13 2113 63
NM_001380339.1 NP_001367466.1 12 - 13 2113 63
GenBank mRNA AF168688.1 - 12 - 13 2179 63
AJ243179.2 - 12 - 13 2201 63
AJ243180.2 - 13 - 14 2274 63
AJ243181.2 - 12 - 13 2194 63
AJ243182.2 - 13 - 14 2228 63

View Full Product Details