Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-73
Entrez Gene ID
188159
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_T06C10.2
Chromosome Location
Chr.IV: 7879317 - 7881055 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068909.3 NP_501310.2 2 - 3 215 153

View Full Product Details