Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
czw-1
Entrez Gene ID
177587
Gene Name
Centromere/kinetochore protein zw10 homolog
Gene Aliases
CELE_F20D12.4
Chromosome Location
Chr.IV: 7943614 - 7946825 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068926.6 NP_501327.1 5 - 6 2317 91

View Full Product Details