Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-24
Entrez Gene ID
177594
Gene Name
PHB domain-containing protein
Gene Aliases
CELE_F57H12.2
Chromosome Location
Chr.IV: 7979870 - 7982011 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001368443.4 NP_001355386.1 3 - 4 410 121

View Full Product Details