Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
isp-1
Entrez Gene ID
177609
Gene Name
Cytochrome b-c1 complex subunit Rieske, mitochondrial
Gene Aliases
CELE_F42G8.12
Chromosome Location
Chr.IV: 8132686 - 8136539 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068960.5 NP_501361.1 5 - 6 665 63

View Full Product Details