Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pmk-3
Entrez Gene ID
177610
Gene Name
protein-coding
Gene Aliases
CELE_F42G8.4
Chromosome Location
Chr.IV: 8139220 - 8143146 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307276.3 NP_001294205.1 1 - 2 67 66
NM_001383136.1 5 - 6 661 66

View Full Product Details