Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-52
Entrez Gene ID
181855
Gene Name
C-type LECtin
Gene Aliases
CELE_B0218.8
Chromosome Location
Chr.IV: 8167164 - 8168813 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068970.6 NP_501371.1 5 - 6 753 86

View Full Product Details