Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ocr-2
Entrez Gene ID
188314
Gene Name
protein-coding
Gene Aliases
CELE_T09A12.3
Chromosome Location
Chr.IV: 8213788 - 8220087 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068979.5 NP_501380.1 5 - 6 1312 78

View Full Product Details