Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-6
Entrez Gene ID
184621
Gene Name
Neur_chan_LBD domain-containing protein
Gene Aliases
CELE_F17E9.8
Chromosome Location
Chr.IV: 8339936 - 8342112 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069009.3 NP_501410.3 7 - 8 991 130

View Full Product Details