Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-7
Entrez Gene ID
260200
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_C28C12.13
Chromosome Location
Chr.IV: 8492542 - 8494520 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069049.2 NP_501450.1 1 - 2 223 116

View Full Product Details