Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-33E2
Entrez Gene ID
185653
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_F42A9.5
Chromosome Location
Chr.IV: 8612334 - 8614695 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069069.5 NP_501470.2 3 - 4 287 106

View Full Product Details