Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-33E3
Entrez Gene ID
185652
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_F42A9.4
Chromosome Location
Chr.IV: 8614995 - 8616023 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069070.2 NP_501471.1 1 - 2 114 112

View Full Product Details