Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hst-1
Entrez Gene ID
177675
Gene Name
Heparan sulfate N-sulfotransferase 1
Gene Aliases
CELE_F08B4.6
Chromosome Location
Chr.IV: 8688271 - 8691513 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069090.5 NP_501491.4 4 - 5 1516 93
GenBank mRNA AB038044.1 - 4 - 5 1536 93

View Full Product Details