Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
grsp-2
Entrez Gene ID
177693
Gene Name
protein-coding
Gene Aliases
CELE_T28C6.1
Chromosome Location
Chr.IV: 8816331 - 8818512 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268403.3 NP_001255332.1 3 - 4 458 74
NM_001268404.3 NP_001255333.1 3 - 4 422 74
NM_001268405.3 NP_001255334.1 3 - 4 435 74
NM_001268406.4 NP_001255335.1 3 - 4 473 74
NM_001268407.3 NP_001255336.1 3 - 4 422 74
NM_001268408.3 NP_001255337.1 2 - 3 218 74
NM_001307124.3 NP_001294053.1 3 - 4 458 74
NM_001372890.2 NP_001360508.1 2 - 2 206 74
NM_001372891.3 NP_001360509.1 3 - 3 223 74

View Full Product Details