Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acc-2
Entrez Gene ID
183758
Gene Name
Acetylcholine-gated chloride channel subunit acc-2
Gene Aliases
CELE_C53D6.3
Chromosome Location
Chr.IV: 9016777 - 9019186 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069166.3 NP_501567.1 5 - 6 602 112

View Full Product Details