Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-11
Entrez Gene ID
177724
Gene Name
protein-coding
Gene Aliases
CELE_ZC410.1
Chromosome Location
Chr.IV: 9070003 - 9073622 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047598.3 NP_001041063.1 3 - 4 237 72
NM_001268430.4 NP_001255359.1 3 - 4 235 72
GenBank mRNA AY305838.1 - 3 - 4 259 72

View Full Product Details