Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-11
Entrez Gene ID
177724
Gene Name
protein-coding
Gene Aliases
CELE_ZC410.1
Chromosome Location
Chr.IV: 9070003 - 9073622 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047598.3 NP_001041063.1 6 - 7 810 108
NM_001047599.4 NP_001041064.2 3 - 4 459 108
NM_001268430.4 NP_001255359.1 6 - 7 808 108
NM_001268431.3 NP_001255360.1 2 - 3 264 108
NR_003464.1 2 - 3 767 108
GenBank mRNA AY305838.1 - 6 - 7 832 108

View Full Product Details