Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-11
Entrez Gene ID
177724
Gene Name
protein-coding
Gene Aliases
CELE_ZC410.1
Chromosome Location
Chr.IV: 9070003 - 9073622 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047598.3 NP_001041063.1 10 - 11 1277 96
NM_001047599.4 NP_001041064.2 7 - 8 926 96
NM_001268430.4 NP_001255359.1 10 - 11 1275 96
NM_001268431.3 NP_001255360.1 6 - 7 731 96
NR_003464.1 6 - 7 1234 96
GenBank mRNA AY305838.1 - 10 - 11 1299 96

View Full Product Details