Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fem-3
Entrez Gene ID
177734
Gene Name
protein-coding
Gene Aliases
CELE_C01F6.4
Chromosome Location
Chr.IV: 9074671 - 9107610 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268436.3 NP_001255365.1 8 - 9 1363 82
NR_056354.1 3 - 4 506 82
GenBank mRNA X64962.1 - - 756 82

View Full Product Details