Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
daf-10
Entrez Gene ID
184883
Gene Name
Intraflagellar transport protein 122 homolog
Gene Aliases
CELE_F23B2.4
Chromosome Location
Chr.IV: 9136933 - 9146211 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069190.4 NP_501591.2 8 - 9 1333 154
GenBank mRNA DQ360810.1 - 8 - 9 1332 154

View Full Product Details