Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-17
Entrez Gene ID
185474
Gene Name
CUTiclin-Like
Gene Aliases
CELE_F38E11.4
Chromosome Location
Chr.IV: 9449125 - 9457501 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069269.5 NP_501670.2 5 - 6 735 148

View Full Product Details