Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-2
Entrez Gene ID
183145
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_C33A12.13
Chromosome Location
Chr.IV: 9486272 - 9487570 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069274.3 NP_501675.1 1 - 2 447 87

View Full Product Details