Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
elo-1
Entrez Gene ID
177787
Gene Name
protein-coding
Gene Aliases
CELE_F56H11.4
Chromosome Location
Chr.IV: 9526366 - 9528350 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276838.3 NP_001263767.1 2 - 3 555 70
NM_069288.8 NP_501689.1 3 - 4 738 70
GenBank mRNA AF244356.1 - 3 - 4 717 70

View Full Product Details