Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spp-18
Entrez Gene ID
185015
Gene Name
protein-coding
Gene Aliases
CELE_F27C8.4
Chromosome Location
Chr.IV: 9593667 - 9594413 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069303.5 NP_501704.1 2 - 3 184 98

View Full Product Details