Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
del-2
Entrez Gene ID
177796
Gene Name
protein-coding
Gene Aliases
CELE_F58G6.6
Chromosome Location
Chr.IV: 9636905 - 9642025 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268482.3 NP_001255411.1 10 - 11 1599 106
NM_001380416.1 NP_001367239.1 6 - 7 1181 106

View Full Product Details