Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
amph-1
Entrez Gene ID
177797
Gene Name
AMPHiphysin homolog
Gene Aliases
CELE_F58G6.1
Chromosome Location
Chr.IV: 9644221 - 9647929 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069310.5 NP_501711.1 2 - 3 252 89

View Full Product Details