Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-38
Entrez Gene ID
191720
Gene Name
protein-coding
Gene Aliases
CELE_K01H12.3
Chromosome Location
Chr.IV: 9710187 - 9712420 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306992.3 NP_001293921.1 3 - 4 271 84
NM_069327.3 NP_501728.2 7 - 8 827 84

View Full Product Details