Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pis-1
Entrez Gene ID
177818
Gene Name
protein-coding
Gene Aliases
CELE_T13F2.3
Chromosome Location
Chr.IV: 9791933 - 9796594 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069347.4 NP_501748.1 12 - 13 2718 177
NM_069348.7 NP_501749.1 12 - 13 2714 177

View Full Product Details