Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-4
Entrez Gene ID
177839
Gene Name
Nuclear hormone receptor family member nhr-4
Gene Aliases
CELE_F32B6.1
Chromosome Location
Chr.IV: 9879017 - 9882154 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069374.10 NP_501775.1 8 - 9 1277 83
GenBank mRNA AF083223.1 - 7 - 8 1194 83

View Full Product Details