Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbc-3
Entrez Gene ID
177845
Gene Name
protein-coding
Gene Aliases
CELE_F32B6.8
Chromosome Location
Chr.IV: 9899012 - 9902513 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027993.4 NP_001023164.1 6 - 7 756 80
NM_001027994.4 NP_001023165.1 7 - 8 829 80

View Full Product Details