Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dis-3
Entrez Gene ID
177875
Gene Name
Probable exosome complex exonuclease RRP44
Gene Aliases
CELE_C04G2.6
Chromosome Location
Chr.IV: 10097196 - 10106523 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069434.5 NP_501835.2 12 - 13 2758 92

View Full Product Details