Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-59
Entrez Gene ID
187799
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_R11A8.3
Chromosome Location
Chr.IV: 10360636 - 10363339 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069510.3 NP_501911.1 4 - 5 1006 128

View Full Product Details