The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_069537.7 | NP_501938.1 | 6 - 7 | 2041 | 62 |
GenBank mRNA | AF038576.1 | - | 6 - 7 | 2062 | 62 |