Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zim-1
Entrez Gene ID
3565259
Gene Name
protein-coding
Gene Aliases
CELE_T07G12.6
Chromosome Location
Chr.IV: 10547086 - 10559960 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136397.4 NP_001129869.1 6 - 7 1653 78
NM_001276847.3 NP_001263776.1 3 - 4 505 78
NM_001276848.3 NP_001263777.1 3 - 4 505 78
NM_069547.4 NP_501948.2 6 - 7 1651 78

View Full Product Details