Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ptr-14
Entrez Gene ID
177951
Gene Name
SSD domain-containing protein
Gene Aliases
CELE_R09H10.4
Chromosome Location
Chr.IV: 10615427 - 10620958 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069562.4 NP_501963.1 5 - 6 1163 67

View Full Product Details