Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ugt-54
Entrez Gene ID
188885
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_T25B9.7
Chromosome Location
Chr.IV: 10761698 - 10764496 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069595.6 NP_501996.2 7 - 8 1086 152

View Full Product Details