Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
miz-1
Entrez Gene ID
3565074
Gene Name
protein-coding
Gene Aliases
CELE_R13.4
Chromosome Location
Chr.IV: 10812407 - 10825979 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268583.2 NP_001255512.1 5 - 6 794 60
NM_001268584.2 NP_001255513.1 1 - 2 272 60
NM_001307057.4 NP_001293986.1 5 - 6 796 60

View Full Product Details