Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- atg-7
- Entrez Gene ID
-
178005
- Gene Name
- Ubiquitin-like modifier-activating enzyme ATG7
- Gene Aliases
-
CELE_M7.5
- Chromosome Location
-
Chr.IV: 11078671 - 11082254 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069663.5 |
NP_502064.1 |
5 - 6 |
1559 |
80 |
- Gene Symbol
- let-70
- Entrez Gene ID
-
178006
- Gene Name
- Ubiquitin-conjugating enzyme E2 2
- Gene Aliases
-
CELE_M7.1
- Chromosome Location
-
Chr.IV: 11078694 - 11082253 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069664.6 |
NP_502065.1 |
7 - 8 |
1467 |
80 |
View Full Product Details