Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
let-70
Entrez Gene ID
178006
Gene Name
Ubiquitin-conjugating enzyme E2 2
Gene Aliases
CELE_M7.1
Chromosome Location
Chr.IV: 11078694 - 11082253 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069664.6 NP_502065.1 3 - 4 345 85

View Full Product Details