Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bcc-1
Entrez Gene ID
178008
Gene Name
SAM domain-containing protein
Gene Aliases
CELE_M7.3
Chromosome Location
Chr.IV: 11086074 - 11092552 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069666.6 NP_502067.2 4 - 5 519 113

View Full Product Details