Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nhr-67
Entrez Gene ID
178024
Gene Name
Nuclear hormone receptor family member nhr-67
Gene Aliases
CELE_C08F8.8
Chromosome Location
Chr.IV: 11169748 - 11174791 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069693.5 NP_502094.1 2 - 3 224 81

View Full Product Details