Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dnj-1
Entrez Gene ID
178040
Gene Name
protein-coding
Gene Aliases
CELE_B0035.14
Chromosome Location
Chr.IV: 11299036 - 11304790 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069721.7 NP_502122.1 5 - 6 934 71

View Full Product Details