Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hum-8
Entrez Gene ID
176872
Gene Name
protein-coding
Gene Aliases
CELE_Y66H1A.6
Chromosome Location
Chr.IV: 429209 - 440554 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028377.5 NP_001023548.2 8 - 9 2061 118
NM_001307645.3 NP_001294574.1 8 - 9 2061 118

View Full Product Details