Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pig-1
Entrez Gene ID
176877
Gene Name
protein-coding
Gene Aliases
CELE_W03G1.6
Chromosome Location
Chr.IV: 506731 - 519457 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028249.6 NP_001023420.2 6 - 7 1038 113
NM_001028250.5 NP_001023421.2 6 - 7 1038 113
NM_001307424.2 NP_001294353.1 6 - 7 1039 113
NM_001307425.1 NP_001294354.1 6 - 7 1038 113
NM_001307428.3 NP_001294357.1 2 - 3 306 113
NM_001307429.3 NP_001294358.1 2 - 3 306 113
NM_001380011.1 NP_001368238.1 2 - 3 306 113
NM_001380012.1 NP_001368239.1 2 - 3 306 113

View Full Product Details