Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
asm-3
Entrez Gene ID
176879
Gene Name
protein-coding
Gene Aliases
CELE_W03G1.7
Chromosome Location
Chr.IV: 520214 - 523809 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047531.2 NP_001040996.1 7 - 8 978 113
NM_001047532.6 NP_001040997.1 6 - 7 776 113

View Full Product Details