Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-76
Entrez Gene ID
182276
Gene Name
Nuclear Hormone Receptor family
Gene Aliases
CELE_C05G6.1
Chromosome Location
Chr.IV: 627537 - 633966 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067560.5 NP_499961.2 1 - 2 76 69

View Full Product Details