Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ech-5
Entrez Gene ID
176908
Gene Name
Enoyl-CoA Hydratase
Gene Aliases
CELE_F56B3.5
Chromosome Location
Chr.IV: 797023 - 799923 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067592.6 NP_499993.2 4 - 5 449 65

View Full Product Details